Skip to Content
Merck
All Photos(2)

Key Documents

ABN2241

Sigma-Aldrich

Anti-GLI2

from rabbit, purified by affinity chromatography

Synonym(s):

Zinc finger protein GLI2, GLI family zinc finger protein 2, Tax helper protein

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€365.00

€365.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
€365.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€365.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... GLI2(2736)
mouse ... Gli2(14633)

General description

Zinc finger protein GLI2 (UniProt: P10070; also known as GLI family zinc finger protein 2, Tax helper protein) is encoded by the Gli2 (also known as THP) gene (Gene ID: 2736) in human. Gli2 belongs to the family of Gli C2H2-type zinc-finger proteins and contains 5 C3H2-type zinc fingers. Its N-terminal domain confers transcriptional repressor activity, while the C-terminal domain mediates transcriptional activation. Transcriptional activity of Gli proteins is also regulated through its binding to Suppressor of Fu (Sufu), which is a negative regulator of Hedgehog signaling in Drosophila as well as in vertebrates. Sufu binds to all three Gli proteins with different affinities. Phosphorylation of Gli2 by DYRK2 is shown to block its transcriptional activity and promotes its proteasomal degradation. Gli2 is reported to be essential for normal embryonic development. Gli2 affects ventroposterior mesodermal development through its regulation of Wnt, Brachyury and Xhox3 gene families. Knockout studies have shown that Gli1 can compensate for Gli2, but replacement is not perfect and is context dependent. Gli2 null mice embryos develop neural tube defects and double knockout Gli1 and Gli2 mice display serious central nervous system and lung defects. Overexpression of Gli2 has been linked to tumorigenesis and can lead to basal cell carcinomas. Mutations in Gli2 gene have been linked to holoprosencephaly 9 that is characterized by failure of forebrain to correctly separate into right and left hemispheres. Gli2 gene mutations are also known to cause Culler-Jones syndrome that features pituitary dysfunction, syndactyly, hypothalamic hamartoma, and visceral malformations.

Specificity

This polyclonal antibody detects Gli2 in mouse brain. It targets an epitope within 12 amino acids from the N-terminal half of isoform 5.

Immunogen

KLH-conjugated linear peptide corresponding to 12 amino acids from the internal edge of N-terminal region of Human Gli2.

Application

Anti-GLI2, Cat. No. ABN2241, is a highly specific rabbit polyclonal antibody that targets Zinc finger protein GLI2 and has been tested in Immunohistochemistry (Paraffin) and Western Blotting.
Immunohistochemistry Analysis: A 1:250 dilution of this antibody detected GLI2 in human pancreas, human cerebral cortex, and rat testis.
Research Category
Neuroscience

Quality

Evaluated by Western Blotting in mouse brain tissue lysate.

Western Blotting Analysis: 2 µg/mL of this antibody detected GLI2 in 10 µg of mouse brain tissue lysate.

Target description

~170 kDa observed; 167.78 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Linkage

Replaces: ABN506

Physical form

Affinity Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Virender Kumar et al.
Biomaterials, 295, 122049-122049 (2023-02-25)
Alcohol-associated liver disease (ALD) and its complications are significant health problems worldwide. Several pathways in ALD are influenced by alcohol that drives inflammation, fatty acid metabolism, and fibrosis. Although miR-96 has become a key regulator in several liver diseases, its

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service