Skip to Content
Merck

Skip To

ABC91

Anti-HUNK Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Hormonally up-regulated neu tumor-associated kinase, B19, Serine/threonine-protein kinase MAK-V

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IHC, WB
Citations:
-


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin), western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... HUNK(30811)

General description

Hormonally up-regulated neu tumor-associated kinase (HUNK) belongs to the protein kinase superfamily and is an SNF1-related serine/threonine kinase that has a unique COOH-terminal (C-terminal) part without homology to any known protein. This protein is highly expressed in the mammary gland, kidney, and central nervous system.
~70 kDa observed. The calculated molecular weight is 80 kDa, however HUNK has been shown as a ~70 kDa band in western blots. (Gardner, H.P., et al. (2000). Development. 127:4493-4509.)

Immunogen

GST-tagged recombinant protein corresponding to human HUNK.

Application

Immunohistochemistry Analysis: A 1:50-1:250 dilution from a representative lot detected HUNK in human brain and human cerebellum tissues.
This Anti-HUNK Antibody is validated for use in WB, IH(P) for the detection of HUNK.

Biochem/physiol Actions

Other homologies: Mouse (87% sequence homology) and Rat (85% sequence homology).

Analysis Note

Control
SK-BR3 cell lysate
Evaluated by Western Blot in SK-BR3 cell lysate.

Western Blot Analysis: 0.5 µg/mL of this antibody detected HUNK on 10 µg of SK-BR3 cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SAB4500475ABS159MABS1882
antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

purified antibody

antibody form

purified immunoglobulin

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

mouse

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

shipped in

ambient

clone

polyclonal

clone

polyclonal

clone

polyclonal

clone

1H9.1, monoclonal


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters