Skip to Content
Merck
All Photos(1)

Key Documents

AB3274

Sigma-Aldrich

Anti-Aquaporin 2 Antibody

Chemicon®, from rabbit

Synonym(s):

AQP2, AQP-CD, WCD-CD

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, human

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... AQP2(359)

Specificity

Recognizes Aquaporin 2.

SPECIES REACTIVITIES: The epitope specific for Aquaporin 2 is identical in rat and mouse and only slightly different in human (17/18 amino acids). Reactivity with other species has not yet been tested.

Immunogen

Highly purified peptide corresponding to residues 254-271 of rat or mouse Aquaporin 2 (SwissProt accession number P34080).

Application

Anti-Aquaporin 2 Antibody is an antibody against Aquaporin 2 for use in WB, IH.
Research Category
Neuroscience
Research Sub Category
Ion Channels & Transporters
Western blotting: 1:200-1:1000 using ECL on rat kidney membranes.

Immunohistochemistry on rat kidney sections.

Dilutions should be made using a carrier protein such as BSA (1-3%).

Optimal working dilutions must be determined by the end user.

Physical form

Affinity purified immunoglobulin. Lyophilized from PBS, pH 7.4, containing 1% BSA and 0.05% sodium azide. Reconstitute with 200 μL of sterile distilled water. Centrifuge antibody preparation before use (10,000 x g for 5 min).

Storage and Stability

Maintain lyophilized material at -20°C for up to 6 months after date of receipt. After reconstitution maintain at -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.

Analysis Note

Control
Included free of charge with the antibody is 40 μg of control antigen (lyophilized powder). Reconstitute with 100 μL of deionized water. For negative control, preincubate 0.5ug of purified peptide with 1 μg of antibody for one hour at room temperature.

Optimal concentrations must be determined by the end user.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tanya M Gwathmey et al.
American journal of physiology. Renal physiology, 299(5), F983-F990 (2010-09-03)
The kidney is an important target for the actions of the renin-angiotensin system (RAS) and this tissue contains a complete local RAS that expresses the bioactive peptides angiotensin II (ANG II) and Ang-(1-7). We find both angiotensin type 1 (AT(1)R)
Stine Julie Tingskov et al.
Frontiers in physiology, 10, 948-948 (2019-08-27)
Arginine vasopressin (AVP) mediates water reabsorption in the kidney collecting ducts through regulation of aquaporin-2 (AQP2). Also, estrogen has been known to regulate AQP2. Consistently, we previously demonstrated that tamoxifen (TAM), a selective estrogen receptor modulator, attenuates the downregulation of
Hye-Young Kim et al.
American journal of physiology. Renal physiology, 296(3), F543-F555 (2009-01-09)
Ammonia metabolism and transport are critical for acid-base homeostasis. The ammonia transporter family member Rh C glycoprotein (Rhcg) is expressed in distal renal tubular segments, and its expression is regulated in parallel with renal ammonia metabolism. However, there are inconsistencies
Hyo-Jung Choi et al.
Cells, 9(5) (2020-05-18)
Sorting nexin 27 (SNX27), a PDZ (Postsynaptic density-95/Discs large/Zonula occludens 1) domain-containing protein, cooperates with a retromer complex, which regulates intracellular trafficking and the abundance of membrane proteins. Since the carboxyl terminus of aquaporin-2 (AQP2c) has a class I PDZ-interacting
Emily S Dorey et al.
Physiological reports, 7(21), e14273-e14273 (2019-11-07)
Maternal alcohol consumption can impair renal development and program kidney dysfunction in offspring. Given that most women who drink alcohol cease consumption upon pregnancy recognition, we aimed to investigate the effect of alcohol around the time of conception (PC:EtOH) on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service