Skip to Content
Merck
All Photos(1)

Key Documents

902267

Supelco

Rapid-action connector

3/8 in. (product side)

Sign Into View Organizational & Contract Pricing

Select a Size

1 UNIT
€614.00

€614.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 UNIT
€614.00

About This Item

UNSPSC Code:
41121815
NACRES:
NB.51

€614.00


Please contact Customer Service for Availability

Request a Bulk Order

fitting

3/8 in. (product side)

shipped in

ambient

storage temp.

no temp limit

General description

(product side) with 3/8" thread
Our withdrawal and safety components are easily interconnectable thanks to a wide selection of reducers, adapters and couplings. Due to the wide variety, many different customized installations are possible – everything for your individual requirements.
This rapid-action connector is made of stainless steel with a special high quality sealing for maximum protection of our solvents quality. It is equipped with coupling (female) Type 21 and G3/8 outer thread.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service