Skip to Content
Merck
All Photos(1)

Key Documents

8.51057

Sigma-Aldrich

Z(2-Cl)-OSu

Novabiochem®

Synonym(s):

Z(2-Cl)-OSu, N-(2-Chlorobenzyloxycarbonyloxy)succinimide

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€20.60
100 G
€43.10

€20.60


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€20.60
100 G
€43.10

About This Item

Empirical Formula (Hill Notation):
C12H10ClNO5
CAS Number:
Molecular Weight:
283.66
MDL number:
UNSPSC Code:
12352108
NACRES:
NA.22

€20.60


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

product line

Novabiochem®

form

powder

potency

>2000 mg/kg LD50, oral (Rat)

manufacturer/tradename

Novabiochem®

mp

101-105 °C

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

Clc1c(cccc1)COC(=O)ON2C(=O)CCC2=O

InChI

1S/C12H10ClNO5/c13-9-4-2-1-3-8(9)7-18-12(17)19-14-10(15)5-6-11(14)16/h1-4H,5-7H2

InChI key

LVZHXYXNMHCBKC-UHFFFAOYSA-N

General description

A reagent for the mild introduction of the Z(2-Cl)-amino protecting group.

Linkage

Replaces: 01-63-0003

Analysis Note

Colour (visual): white to yellow to beige
Appearance of substance (visual): powder
Melting range (lower value): ≥ 98 °C
Melting range (upper value): ≤ 104 °C

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service