Skip to Content
Merck
All Photos(1)

Documents

8.41834

Sigma-Aldrich

4-Benzyloxyphenol

for synthesis

Synonym(s):

4-Benzyloxyphenol, Hydroquinone monobenzyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H12O2
CAS Number:
Molecular Weight:
200.23
MDL number:
UNSPSC Code:
12352104
EC Index Number:
203-083-3

Quality Level

Assay

≥98.0% (HPLC)

form

solid

mp

117.9-121.5 °C

density

1.26 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C13H12O2/c14-12-6-8-13(9-7-12)15-10-11-4-2-1-3-5-11/h1-9,14H,10H2

InChI key

VYQNWZOUAUKGHI-UHFFFAOYSA-N

Analysis Note

Assay (HPLC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 118 °C
Melting range (upper value): ≤ 121 °C
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service