Skip to Content
Merck
All Photos(1)

Key Documents

8.41642

Sigma-Aldrich

4-Bromo-1-butene

for synthesis

Synonym(s):

4-Bromo-1-butene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H7Br
CAS Number:
Molecular Weight:
135.00
MDL number:
UNSPSC Code:
12352101
EC Index Number:
225-937-4

Quality Level

Assay

≥97.0% (GC)

form

liquid

transition temp

flash point 1 °C

solubility

0.76 g/L

density

1.33 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C4H7Br/c1-2-3-4-5/h2H,1,3-4H2

InChI key

DMAYBPBPEUFIHJ-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 97.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.330 - 1.340
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Resp. Sens. 1 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(C)

1 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service