Skip to Content
Merck
All Photos(1)

Key Documents

8.21959

Sigma-Aldrich

Benzenesulfonamide

for synthesis

Synonym(s):

Benzenesulfonamide

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€35.70
500 G
€71.00

€35.70


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 G
€35.70
500 G
€71.00

About This Item

Empirical Formula (Hill Notation):
C6H7NO2S
CAS Number:
Molecular Weight:
157.19
MDL number:
UNSPSC Code:
12352111
EC Index Number:
202-637-1
NACRES:
NA.22

€35.70


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98.0% (HPLC)

form

powder

potency

991 mg/kg LD50, oral (Rat)

mp

149-152 °C

transition temp

flash point 250 °C

solubility

4.3 g/L

bulk density

700 kg/m3

storage temp.

2-30°C

SMILES string

[S](=O)(=O)(N)c1ccccc1

InChI

1S/C6H7NO2S/c7-10(8,9)6-4-2-1-3-5-6/h1-5H,(H2,7,8,9)

InChI key

KHBQMWCZKVMBLN-UHFFFAOYSA-N

Analysis Note

Assay (HPLC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 150 °C
Melting range (upper value): ≤ 155 °C
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service