Skip to Content
Merck
All Photos(1)

Key Documents

8.21112

Sigma-Aldrich

N1-2-Thiazolylsulfanilamid

for synthesis

Synonym(s):

N1-2-Thiazolylsulfanilamid, Sulfathiazol

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€34.50
250 G
€169.00

€34.50


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€34.50
250 G
€169.00

About This Item

Empirical Formula (Hill Notation):
C9H9N3O2S2
CAS Number:
Molecular Weight:
255.32
MDL number:
UNSPSC Code:
12352111
EC Index Number:
200-771-5
NACRES:
NA.22

€34.50


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

form

powder

mp

199-202 °C

storage temp.

no temp limit

SMILES string

[S](=O)(=O)(Nc2[s]ccn2)c1ccc(cc1)N

InChI

1S/C9H9N3O2S2/c10-7-1-3-8(4-2-7)16(13,14)12-9-11-5-6-15-9/h1-6H,10H2,(H,11,12)

InChI key

JNMRHUJNCSQMMB-UHFFFAOYSA-N

Analysis Note

Assay (HClO₄): ≥ 98.0 %
Melting range (lower value): ≥ 199 °C
Melting range (upper value): ≤ 202 °C
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service