Skip to Content
Merck
All Photos(1)

Key Documents

8.20869

Sigma-Aldrich

2-Naphthyl phenyl ketone

for synthesis

Synonym(s):

2-Naphthyl phenyl ketone, Phenyl 2-naphthyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H12O
CAS Number:
Molecular Weight:
232.28
UNSPSC Code:
12352115
EC Index Number:
211-410-6
NACRES:
NA.22

Quality Level

Assay

≥98.0% (HPLC)

form

powder

bp

398 °C

mp

80-82 °C

storage temp.

2-30°C

InChI

1S/C17H12O/c18-17(14-7-2-1-3-8-14)16-11-10-13-6-4-5-9-15(13)12-16/h1-12H

InChI key

SJNXJRVDSTZUFB-UHFFFAOYSA-N

Analysis Note

Assay (HPLC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 78 °C
Melting range (upper value): ≤ 82 °C
Identity (IR): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service