Skip to Content
Merck
All Photos(1)

Key Documents

8.18804

Sigma-Aldrich

Trimethyl orthoacetate

for synthesis

Synonym(s):

Trimethyl orthoacetate, 1,1,1-Trimethoxyethane, Orthoacetic acid trimethyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€89.30
500 ML
€175.00

€89.30


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
€89.30
500 ML
€175.00

About This Item

Linear Formula:
CH3C(OCH3)3
CAS Number:
Molecular Weight:
120.15
MDL number:
UNSPSC Code:
12352112
EC Index Number:
215-892-9
NACRES:
NA.22

€89.30


Please contact Customer Service for Availability

Request a Bulk Order

vapor pressure

20 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

potency

>2000 mg/kg LD50, oral (Rat)

bp

108 °C/1013 hPa

transition temp

flash point 15 °C

density

0.96 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O(C(OC)(OC)C)C

InChI

1S/C5H12O3/c1-5(6-2,7-3)8-4/h1-4H3

InChI key

HDPNBNXLBDFELL-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.956 - 0.958
Identity (IR): passes test

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Sens. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service