Skip to Content
Merck
All Photos(1)

Documents

8.18424

Sigma-Aldrich

Ethylhexadecyldimethylammonium bromide

for synthesis

Synonym(s):

Ethylhexadecyldimethylammonium bromide, Hexadecylethyldimethylammonium bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5N(CH3)2C16H33Br
CAS Number:
Molecular Weight:
378.47
MDL number:
UNSPSC Code:
12352107
EC Index Number:
204-672-8
NACRES:
NA.22

Quality Level

form

solid

potency

500 mg/kg LD50, oral (Rat)

mp

178-186 °C

storage temp.

2-30°C

InChI

1S/C20H44N.BrH/c1-5-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21(3,4)6-2;/h5-20H2,1-4H3;1H/q+1;/p-1

InChI key

VUFOSBDICLTFMS-UHFFFAOYSA-M

Application

  • Heavy metal removal from industrial effluents: Ethylhexadecyldimethylammonium bromide is utilized as an ionic solid impregnated in chitosan for the effective removal of heavy metals like Cd(II) and Hg(II) from industrial effluents. This application underscores its potential in environmental cleanup and wastewater treatment technologies (Shekhawat et al., 2017).

Analysis Note

Assay (argentometric): ≥ 98.0 %
Identity (IR): passes test

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service