Skip to Content
Merck
All Photos(1)

Documents

8.14691

Sigma-Aldrich

2,5-Dimethylresorcinol

for synthesis

Synonym(s):

2,5-Dimethylresorcinol, ß-Orcinol, Dihydroxycylene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H10O2
CAS Number:
Molecular Weight:
138.16
MDL number:
UNSPSC Code:
12352104
EC Index Number:
207-688-3
NACRES:
NA.22

Quality Level

Assay

≥98.0% (HPLC)

form

powder

bp

277-280 °C

mp

161-164 °C

storage temp.

2-30°C

InChI

1S/C8H10O2/c1-5-3-7(9)6(2)8(10)4-5/h3-4,9-10H,1-2H3

InChI key

GHVHDYYKJYXFGU-UHFFFAOYSA-N

Analysis Note

Assay (HPLC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 161 °C
Melting range (upper value): ≤ 164 °C
Identity (IR): passes test

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Eye Dam. 1 - Skin Corr. 1C - Skin Sens. 1B

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service