Skip to Content
Merck
All Photos(1)

Key Documents

8.08529

Sigma-Aldrich

Pyrocatechol dimethyl ether

for synthesis

Synonym(s):

Pyrocatechol dimethyl ether, Veratrole, 1,2-Dimethoxybenzene

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
€52.30
1 L
€108.00

€52.30


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
250 ML
€52.30
1 L
€108.00

About This Item

Linear Formula:
(CH3O)2C6H4
CAS Number:
Molecular Weight:
138.16
MDL number:
UNSPSC Code:
12352112
EC Index Number:
202-045-3
NACRES:
NA.22

€52.30


Please contact Customer Service for Availability

Request a Bulk Order

vapor pressure

0.63 hPa ( 25 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

potency

890 mg/kg LD50, oral (Rat)

mp

22-23 °C

transition temp

flash point 87 °C

solubility

insoluble 6.69 g/L

density

1.085 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O(C)c1c(cccc1)OC

InChI

1S/C8H10O2/c1-9-7-5-3-4-6-8(7)10-2/h3-6H,1-2H3

InChI key

ABDKAPXRBAPSQN-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.084 - 1.086
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

161.6 °F - closed cup

Flash Point(C)

72 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service