Skip to Content
Merck
All Photos(1)

Key Documents

8.02295

Sigma-Aldrich

4-Chloro-3-methylphenol

for synthesis

Synonym(s):

4-Chloro-3-methylphenol, 4-Chloro-m-cresol, 2-Chloro-5-hydroxytoluene

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€30.40
250 G
€40.80
1 KG
€120.00
50 KG
€2,600.00

€30.40


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€30.40
250 G
€40.80
1 KG
€120.00
50 KG
€2,600.00

About This Item

Linear Formula:
4-(Cl)-3-(CH3)C6H3OH
CAS Number:
Molecular Weight:
142.58
MDL number:
UNSPSC Code:
12352104
EC Index Number:
200-431-6
NACRES:
NA.22

€30.40


Please contact Customer Service for Availability

Request a Bulk Order

vapor pressure

<0.1 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

form

powder

autoignition temp.

590 °C

potency

1830 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rat)

pH

6.5 (20 °C, 1 g/L in H2O)

bp

235-239 °C/1013 hPa

mp

63-65 °C

transition temp

flash point 118 °C

solubility

3.9 g/L

density

1.37 g/cm3 at 20 °C

bulk density

800 kg/m3

storage temp.

2-30°C

SMILES string

Clc1c(cc(cc1)O)C

InChI

1S/C7H7ClO/c1-5-4-6(9)2-3-7(5)8/h2-4,9H,1H3

InChI key

CFKMVGJGLGKFKI-UHFFFAOYSA-N

Application

  • Compression of mono-4 chloro, 3-methylphenyl phosphate ester and di-4 chloro, 3-methylphenyl phosphate ester through kinetic study: This study discusses a method using 4-chloro-3-methylphenol and POCl3 taken in a 2:1 ratio, exploring their kinetic behaviors (AR Teli, AA Khan, SH Dar, A Rashid, 2017).
  • Commercially Important Chlorinated Phenols: Reviews the usage of 4-chloro-3-methylphenol among other chlorinated phenols in various industrial applications (K Smith, G El-Hiti).

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Melting range (lower value): ≥ 64 °C
Melting range (upper value): ≤ 67 °C
Identity (IR): passes test

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1C - Skin Sens. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

244.4 °F - closed cup

Flash Point(C)

118.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service