Skip to Content
Merck
All Photos(1)

Documents

8.02169

Sigma-Aldrich

Decanoic acid

for synthesis

Synonym(s):

Decanoic acid, Capric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)8COOH
CAS Number:
Molecular Weight:
172.26
MDL number:
UNSPSC Code:
12352106
EC Index Number:
206-376-4
NACRES:
NA.22

vapor pressure

0.13 hPa ( 79 °C)

Quality Level

Assay

≥98.0% (GC)

potency

>10000 mg/kg LD50, oral (Rat)

pH

4 (20 °C, 0.2 g/L in H2O)

mp

29-32 °C

transition temp

flash point 147 °C

density

0.89 g/cm3 at 20 °C

bulk density

690 kg/m3

storage temp.

2-30°C

InChI

1S/C10H20O2/c1-2-3-4-5-6-7-8-9-10(11)12/h2-9H2,1H3,(H,11,12)

InChI key

GHVNFZFCNZKVNT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 29 °C
Melting range (upper value): ≤ 33 °C
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

296.6 °F - closed cup

Flash Point(C)

147 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service