Skip to Content
Merck
All Photos(1)

Key Documents

8.00920

Sigma-Aldrich

Dicyclohexyl phthalate

for synthesis

Synonym(s):

Dicyclohexyl phthalate, Phthalic acid dicyclohexyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4-1,2-(COOC6H11)2
CAS Number:
Molecular Weight:
330.42
MDL number:
UNSPSC Code:
12352108
EC Index Number:
201-545-9
To inquire about this product 8.00920, please contact your local Merck office or dealer. Contact Technical Service

vapor pressure

0.0000039 hPa ( 50 °C)

Assay

≥99.0% (GC)

form

crystals

autoignition temp.

395 °C

potency

>2000 mg/kg LD50, oral (Rat)

mp

65.6 °C

transition temp

flash point 180-190 °C

solubility

1.015 g/L

density

1.15 g/cm3 at 20 °C

bulk density

710 kg/m3

storage temp.

2-30°C

InChI

1S/C20H26O4/c21-19(23-15-9-3-1-4-10-15)17-13-7-8-14-18(17)20(22)24-16-11-5-2-6-12-16/h7-8,13-16H,1-6,9-12H2

InChI key

VOWAEIGWURALJQ-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Melting range (lower value): ≥ 62 °C
Melting range (upper value): ≤ 64 °C
Identity (IR): passes test

Pictograms

Exclamation markHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Repr. 1B - Skin Sens. 1

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(C)

180 - 190 °C


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service