Skip to Content
Merck
All Photos(1)

Key Documents

553850

Sigma-Aldrich

Anti-Relaxin 2 Rabbit pAb

lyophilized, Calbiochem®

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€497.00

€497.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
€497.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€497.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized

does not contain

preservative

species reactivity

human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

isotype

IgG

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... RLN2(6019)

General description

Purified rabbit polyclonal antibody. Recognizes the relaxin-2 protein.
Recognizes the relaxin-2 protein.
This Anti-Relaxin 2 Rabbit pAb is validated for use in ELISA, Immunohistochemistry, Radioimmunoassay for the detection of Relaxin 2.

Immunogen

Human
purified, human relaxin-2

Application

ELISA (1 μg/ml)

Immunohistochemistry (5 μg/ml)

Radioimmunoassay (1 μg/ml)

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from 50 mM Tris, pH 7.5.

Reconstitution

Reconstitute with 100 µl of sterile distilled H₂O. Following reconstitution, aliquot and freeze (-20°C). Avoid freeze/thaw cycles of solution.

Other Notes

Armbruster, F.P., et al. 2001. Eur. J. Med. Res.6, 1.
Does not cross react with insulin, Zn2+-insulin, IGF-1, IGF-2, spermolaxin, inhibin α-subunit, inhibin, seminal-plasma-inhibin-like peptide, hCG, LH, FSH, prolactin, or ubiquitin. Variables associated with assay conditions will dictate the optimal working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service