Skip to Content
Merck
All Photos(1)

Key Documents

545068

Millipore

Pyridoxamine, Dihydrochloride

Synonym(s):

Pyridoxamine, Dihydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€281.00

€281.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€281.00

About This Item

Empirical Formula (Hill Notation):
C8H12N2O2 · 2HCl · xH2O
CAS Number:
Molecular Weight:
241.11 (anhydrous basis)
MDL number:
UNSPSC Code:
12352116
NACRES:
NA.21

€281.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥99% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

water: 100 mg/mL

shipped in

ambient

storage temp.

10-30°C

SMILES string

[Cl-].[Cl-].[N+H3]Cc1c(c([n+H]cc1CO)C)O

InChI

1S/C8H12N2O2.2ClH/c1-5-8(12)7(2-9)6(4-11)3-10-5;;/h3,11-12H,2,4,9H2,1H3;2*1H

InChI key

HNWCOANXZNKMLR-UHFFFAOYSA-N

Warning

Toxicity: Irritant (B)

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service