Skip to Content
Merck
All Photos(1)

Key Documents

5.34331

Sigma-Aldrich

Hypochlorous Acid Fluorescent Probe, PIS

Synonym(s):

Hypochlorous Acid Fluorescent Probe, PIS, 1,3-Dimethyl-1H-imidazo[4,5-b]phenazine-2(3H)-thione, Hypochlorite Fluorescent Probe, MPO Activity Probe

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H12N4S
Molecular Weight:
280.35
UNSPSC Code:
12352202

Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

brown

solubility

DMF: 0.5 mg/mL (Small yellow particles may be visible.)
DMSO: 0.5 mg/mL

fluorescence

λex ~378 nm
λem ~503 nm

storage temp.

2-8°C

General description

A cell-permeable imidazoline-2-thione based compound that acts as a direct, rapid, selective and sensitive fluorescence turn-on detection probe for hypochlorous acid (HOCl; detection limit of 71 nM). Efficiently reacts with hypochlorite to yield fluorescent imidazolium ions, with good water solubility and photostability. Generates negligible fluorescence emission with several other ROS/RNS (H2O2, TBHP, NO•, ROO•, ONOO- and HO•. Shown to efficiently visualize and detect stimulants triggered endogenous HOCl generation in RAW 264.7 macrophages co-cultured with HeLa cells. Dose-dependently images HOCl levels by two-photon fluorescence microscopy in RAW 264.7 cells and in rat hippocampal slices (depth of >100 µm).
A cell-permeable imidazoline-2-thione based compound that acts as a direct, rapid, selective and sensitive fluorescence turn-on detection probe for hypochlorous acid (HOCl; detection limit of 71 nM). Efficiently reacts with hypochlorite to yield fluorescent imidazolium ions, with good water solubility and photostability. Generates negligible fluorescence emission with several other ROS/RNS (H2O2, TBHP, NO·, ROO·, ONOO and HO·). Shown to efficiently visualize and detect stimulants triggered endogenous HOCl generation in RAW 264.7 macrophages co-cultured with HeLa cells. Also enables to monitor HOCl levels by two-photon image in RAW 264.7 cells and in rat hippocampal slices (depth of >100 µm) in a dose-dependent manner.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
hypochlorous acid

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Xu, Q., et al. 2015. Angew. Chem. Int. Ed.54, 4890.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service