Skip to Content
Merck
All Photos(1)

Key Documents

5.32729

Sigma-Aldrich

RIP1 Kinase Inhibitor III

Synonym(s):

RIP1 Kinase Inhibitor III, 1-(4-(4-Aminofuro[2,3-d]pyrimidin-5-yl)phenyl)-3-(2-fluoro-5-(trifluoromethyl)phenyl)urea, RIP1 Inhibitor III, Receptor-Interacting Protein 1 Inhibitor III

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H13F4N5O2
CAS Number:
Molecular Weight:
431.34
UNSPSC Code:
12352200

Assay

≥98% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

light orange

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

General description

A cell-permeable furopyrimidinyl-phenyl urea compound that potently inhibits RIP1 kinase activity (IC50 = 13 nM; [ATP] = 50 µM) by targeting RIPK1 ATP-binding pocket in its inactive DLG-out conformation (IC50 = 63 nM in competitive binding assays against 5 nM ATP-site ligand) without affecting RIPK2 or RIPK5 activity even at concentrations as high as 1 µM ([ATP] = 10 µM). Effectively prevents TNFα-induced necroptosis in Z-VAD(OMe)-FMK- (Cat. No. 627610) treated U937 cultures (EC50 = 250 nM) in vitro and is efficacious in protecting mice from body temperature loss (by 77%; 20 mg/kg p.o.) when administered 15 min prior to TNFα i.v. injection in vivo with good oral availability (plasma Cmax = 1100 ng/mL, Tmax = 4 h, AUC = 14 µg·h/mL; 2 mg/kg p.o.). Selectivity profiling in a 300-kinase panel shows significant inhibition against only 25 targets (IC50<1 µM), notably RET, EPHA7, FLT3, EPHA6, HIPK4, DDR2, FMS, PDGFRa, and TrkC (% inhibition at 1 µM = 97%, 97%, 96%, 92%, 91%, 90%, 86%, 84%, 84%, respectively).
A cell-permeable, orally available furopyrimidinyl-phenyl urea compound that acts as a potent and reversible inhibitor of receptor interacting protein 1 (RIP1) kinase (IC50 = 63 nM and 13 nM in RIP fluorescence polarization and ADP-Glo kinase assays, respectively). Binds to the ATP-binding pocket of RIP1 kinase with enzyme adopting a DLG-out inactive conformation. Shown to be moderately effective in inhibiting 25 other protein kinases in a screening of 300 kinases by radiolabeled assay, but only at high concentrations (~ 1 µM in the presence of 10 mM ATP). Blocks TNFα-induced necrotic cell death (IC50 = 250 nM in U937 cells) and protects mice from TNFα-induced hypothermic shock when injected 15 min. prior to i.v. administration of TNFα. Displays desirable pharmacokinetic properties with good systemic exposure following an oral dose (AUC= 14 mg/h/ml; Cmax = 1,100 ng/ml at 4 hours).

Please note that the molecular weight for this compound is batch-specific due to variable water content.
A cell-permeable, potent, reversible inhibitor of RIP1 kinase (IC₅₀ = 63 nM in RIP fluorescence polarization assay).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
RIP1 Kinase

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C).
Use only fresh DMSO for reconstitution.

Other Notes

Harris, P.A., et al. 2013. ACS Med. Chem. Lett.4, 1238.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service