Skip to Content
Merck
All Photos(1)

Key Documents

5.09166

Sigma-Aldrich

Windorphen, Inactive Control

Synonym(s):

Windorphen, Inactive Control

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H15ClO3
CAS Number:
Molecular Weight:
302.75
UNSPSC Code:
12352200

Assay

≥98% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

light beige

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

InChI

1S/C17H15ClO3/c1-20-14-7-3-12(4-8-14)16(11-19)17(18)13-5-9-15(21-2)10-6-13/h3-11H,1-2H3/b17-16-

InChI key

VNRALGZMXHFBPG-MSUUIHNZSA-N

General description

(E)-Isomer of 3-Chloro-2,3-bis(4-methoxyphenyl)acrylaldehyde that serves as a suitable inactive control for p300/CBP Inhibitor X, Windorphen (Cat. no. 509164).

Please note that the molecular weight for this compound is batch-specific due to variable water content.

Biochem/physiol Actions

Cell permeable: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 month at -20°C.
Use only fresh DMSO for reconstitution.

Other Notes

Hao. J., et al. 2013 Cell Rep.4, 898.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service