Skip to Content
Merck

5.00112

SAFC

Cocarboxylase chloride

thiamine diphosphoric acid ester chloride EMPROVE® API

Pharma Manufacturing

Synonym(s):

Cocarboxylase chloride, Thiaminium diphosphoric acid ester chloride, Thiaminium pyrophosphate chloride, Thiaminium pyrophosphoric acid ester chloride, TPP

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
Inquire
1 KG
Inquire
5 KG
Inquire

This item is not able to be purchased as a guest user. For further information, please contact us at +34 912 158 016or

.


Select a Size

Change View
100 G
Inquire
1 KG
Inquire
5 KG
Inquire

About This Item

CAS Number:
Molecular Weight:
460.77
MDL number:
UNSPSC Code:
12352107
EC Index Number:
205-836-1

This item is not able to be purchased as a guest user. For further information, please contact us at +34 912 158 016or

.

Quality Level

product line

EMPROVE® API

form

solid

pH

1.0-1.4 (20 °C, 100 g/L in H2O)

mp

240-244 °C (decomposition)

solubility

220 g/L

bulk density

500 kg/m3

application(s)

liquid formulation
pharmaceutical
semi-solid formulation
solid formulation

storage temp.

2-25°C

SMILES string

[P](=O)([O-])(O[P](=O)(O)O)OCCc1[s]c[n+](c1C)Cc2c(nc(nc2)C)N.Cl

InChI

1S/C12H18N4O7P2S.ClH/c1-8-11(3-4-22-25(20,21)23-24(17,18)19)26-7-16(8)6-10-5-14-9(2)15-12(10)13;/h5,7H,3-4,6H2,1-2H3,(H4-,13,14,15,17,18,19,20,21);1H

InChI key

YXVCLPJQTZXJLH-UHFFFAOYSA-N

General description

We provide industry-leading production know-how and comprehensive regulatory support for high-quality APIs, backed by nearly 200 years of experience. From a complete portfolio of innovative folates to custom synthesis, our customized and scalable solutions meet your specific production needs. As part of our EMPROVE® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Cocarboxylase chloride Emprove® API is a source for Vitamin B1 for use in clinical nutrition, vitamin supplements and infusion for Vitamin B1 deficiency. Cocarboxylase Chloride (thiamine diphosphoric acid ester chloride) Emprove® API is manufactured according to ICH Q7 guidelines. An ASMF is available.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


What is the Emprove® Program?

The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:

Elemental Impurities Information

Available by subscription or for a fee

API Information Package

Free of charge - Sign in to download

Learn more about the benefits an Emprove® Program subscription can provide
Why are these documents unavailable?
Emprove® Dossiers contain information intended to support the registration process and risk assessment activities of pharmaceutical producers and contain confidential information. Please contact us if you would like access.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service