Skip to Content
Merck
All Photos(1)

Key Documents

487950

Sigma-Aldrich

NOC-5

A nitric oxide (NO) donor.

Synonym(s):

NOC-5, 3-[2-Hydroxy-1-(1-methylethyl)-2-nitrosohydrazino]-1-propanamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H16N4O2
CAS Number:
Molecular Weight:
176.22
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

white

solubility

0.1 M NaOH: soluble
water: soluble

shipped in

ambient

storage temp.

−20°C

InChI

1S/C6H16N4O2/c1-6(2)9(5-3-4-7)10(12)8-11/h6,12H,3-5,7H2,1-2H3

InChI key

WEHZRCACNLQADC-UHFFFAOYSA-N

General description

A nitric oxide (NO) donor. A stable NO-amine complex that can release two equivalents of NO spontaneously in solution under physiological conditions without a cofactor. Half-life (t½) of NO release is 93 min (PBS, pH 7.4, 22°C).
A nitric oxide (NO) donor. A stable NO-amine complex that can spontaneously release two equivalents of NO in solution under physiological conditions without any cofactor. Half-life (t½) in PBS, pH 7.4 is 93 minutes at 22°C. Relatively stable in alkaline solution (pH ≥10.0).

Biochem/physiol Actions

Cell permeable: no
Primary Target
A nitric oxide (NO) donor
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Irritant (B)

Reconstitution

Following reconstitution in alkaline solution, aliquot and freeze (-20°C). Unstable in aqueous solutions; reconstitute in H₂O just prior to use. Alkaline stock solutions are stable for up to 3 months at -20°C.

Other Notes

Hrabie, J.A., et al. 1993. J. Org. Chem. 58, 1472.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Self-react. C

Storage Class Code

5.2 - Organic peroxides and self-reacting hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service