Skip to Content
Merck
All Photos(1)

Documents

444287

Sigma-Aldrich

MMP-13, Human, Recombinant, Active

Synonym(s):

Matrix metalloproteinase 13

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.77

form

liquid

Quality Level

specific activity

≥50 mU/mg protein

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

shipped in

wet ice

storage temp.

−70°C

General description

Recombinant, human MMP-13. MMP-13 is a collagenase-3 enzyme that degrades a range of extracellular matrix proteins, including collagen types I, II, III, IV, IX, X, and XIV, gelatin, aggrecan, perlecan and fibronectin. It is distinguished from other human collagenases by the fact that it effectively degrades type II collagen. The importance of MMP-13 is further strengthened by reports that it is a biomarker for poor prognosis in many carcinomas and other type of cancers.

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit is defined as the amount of APMA-activated enzyme that will hydrolyze 1µmol MCA-Pro-Leu-Lys-Gly-Leu-DPA-Ala-Arg-NH₂ (Cat. No. 03-32-5032) at 37°C, pH 7.5.

Physical form

In 50 mM HEPES, 10 mM CaCl₂, 20% glycerol, 0.005% BRIJ® 35 Detergent, pH 7.5. APMA-free.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C).

Other Notes

Freije, M.P.S., et al. 1994. J. Biol. Chem.269,16766.

Legal Information

Brij is a registered trademark of Croda International PLC
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service