Skip to Content
Merck
All Photos(1)

Documents

444219-M

Millipore

MMP-1 Substrate III, Fluorogenic

An excellent quenched fluorescence substrate for MMP-1.

Synonym(s):

MMP-1 Substrate III, Fluorogenic

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C51H72N14O12S
Molecular Weight:
1105.27
UNSPSC Code:
12352200

Quality Level

Assay

≥94% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

50% acetic acid: soluble

fluorescence

λex ~365 nm
λem ~450 nm

storage temp.

−20°C

General description

An excellent quenched fluorescence substrate for MMP-1. Has a better kcat/Km value than the MMP Substrate (Cat. No. 444207).
Quenched fluorescence substrate for MMP-1 (excitation maximum: 365 nm; emission maximum: 450 nm). Has a better kcat/Km value than the MMP-Substrate (Cat. No. 444207).

Biochem/physiol Actions

Primary Target
Quenched fluorescence substrate for MMP-1

Warning

Toxicity: Standard Handling (A)

Sequence

DNP-Pro-Cha-Abu-Cys(Me)-His-Ala-Lys(N-Me-Abz)-NH₂[Cha = β-cyclohexylalanyl; Abu = L-α-aminobutyryl; Abz = 2-aminobenzoyl]

Physical form

Supplied as a trifluoroacetate salt.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

McGeehan, G.M., et al. 1994. J. Biol. Chem.269, 32814.
Green, M., et al. 1992. Medicinal Chemistry Annual Meeting, poster (Abstr. 41).

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service