Skip to Content
Merck
All Photos(1)

Key Documents

420130

Millipore

JNK Inhibitor III, Cell-Permeable

The JNK Inhibitor III, Cell-Permeable controls the biological activity of JNK. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

JNK Inhibitor III, Cell-Permeable, HIV-TAT₄₇₋₅₇-gaba-c-Junδ₃₃₋₅₇, Ac-YGRKKRRQRRR-gaba-ILKQSMTLNLADPVGSLKPHLRAKN-NH₂, SAPK Inhibitor III, SAPK Inhibitor III, HIV-TAT₄₇₋₅₇-gaba-c-Junδ₃₃₋₅₇, Ac-YGRKKRRQRRR-gaba-ILKQSMTLNLADPVGSLKPHLRAKN-NH₂

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€268.00

€268.00


Please contact Customer Service for Availability


Select a Size

Change View
1 MG
€268.00

About This Item

Empirical Formula (Hill Notation):
C191H336N70O48S
Molecular Weight:
4413.22
UNSPSC Code:
12352202
NACRES:
NA.77

€268.00


Please contact Customer Service for Availability

Quality Level

Assay

≥95% (HPLC)

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

white

solubility

DMSO: 100 mg/mL
acetic acid: 100 mg/mL

shipped in

wet ice

storage temp.

−20°C

General description

A cell-permeable 37 amino acid peptide constructed by fusing the JNK binding domain sequence (δ) (amino acids 33-57) of human c-Jun to the HIV-TAT transduction domain sequence (amino acids 47-57) with a γ-aminobutyric acid (GABA) spacer. Reported to specifically disrupt c-Jun/JNK complex formation and subsequent phosphorylation and activation of c-Jun by JNK in vitro and in intact cells. Also reported to induce apoptosis in HeLa cells. Mode of inhibition is distinct from that of JNK Inhibitor II (SP600125; Cat. No. 420119), so may complement this inhibitor in JNK pathway studies.
A cell-permeable 37-mer peptide constructed by fusing human c-Jun δ domain (amino acids 33-57) sequence with that of HIV-TAT protein transduction domain (amino acids 47-57) via a γ-aminobutyric acid (GABA) spacer. Shown to specifically disrupt c-Jun/JNK complex formation and the subsequent phosphorylation and activation of c-Jun by JNK both in vitro and in intact cells. Since its mode of inhibition is different than that of JNK Inhibitor II (SP600125; Cat. No. 420119), these two inhibitors can complement each other in JNK signaling pathway studies.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
JNK
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Carcinogenic / Teratogenic (D)

Sequence

Ac-Tyr-Gly-Arg-Lys-Lys-Arg-Arg-Gln-Arg-Arg-Arg-gaba-Ile-Leu-Lys-Gln-Ser-Met-Thr-Leu-Asn-Leu-Ala-Asp-Pro-Val-Gly-Ser-Leu-Lys-Pro-His-Leu-Arg-Ala-Lys-Asn-NH₂

Physical form

Supplied as a trifluoroacetate salt.

Reconstitution

Following reconstitution aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Holzberg, D., et al. 2003. J. Biol. Chem.278, 40213.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service