Skip to Content

368

Trypan Blue Powder

HARLECO® Biological stain designed for connective tissue

Synonym(s):

Trypan Blue, Direct Blue 14, AzidinBlue, Direct blue 14

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C34H24N6O14S4Na4
CAS Number:
Molecular Weight:
960.81
UNSPSC Code:
41116130
EC Index Number:
200-786-7
MDL number:
Beilstein/REAXYS Number:
4360496
Colour Index Number:
23850
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

pH

9.8 (20 °C, 10 g/L in H2O)

mp

>300 °C (lit.)

solubility

10 g/L

shipped in

ambient

SMILES string

[Na+].[Na+].[Na+].[Na+].Cc1cc(ccc1N=Nc2c(O)c3c(N)cc(cc3cc2S([O-])(=O)=O)S([O-])(=O)=O)-c4ccc(N=Nc5c(O)c6c(N)cc(cc6cc5S([O-])(=O)=O)S([O-])(=O)=O)c(C)c4

InChI

1S/C34H28N6O14S4.4Na/c1-15-7-17(3-5-25(15)37-39-31-27(57(49,50)51)11-19-9-21(55(43,44)45)13-23(35)29(19)33(31)41)18-4-6-26(16(2)8-18)38-40-32-28(58(52,53)54)12-20-10-22(56(46,47)48)14-24(36)30(20)34(32)42;;;;/h3-14,41-42H,35-36H2,1-2H3,(H,43,44,45)(H,46,47,48)(H,49,50,51)(H,52,53,54);;;;/q;4*+1/p-4

InChI key

GLNADSQYFUSGOU-UHFFFAOYSA-J

General description

HARLECO® Trypan Blue
368-12 25g Glass Bottle

Application

For In-Vitro Diagnostic Use.
For Professional Use.
Biological stain designed for connective tissue.
HARLECO® Biological stain designed for connective tissue

Legal Information

HARLECO is a registered trademark of Merck KGaA, Darmstadt, Germany


Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library