Skip to Content
Merck
All Photos(1)

Documents

362339

Sigma-Aldrich

Glyco-SNAP-2

A water-soluble nitric oxide (NO) donor that exhibits greater stability (t1/2 = 27 hours) than that of SNAP.

Synonym(s):

Glyco-SNAP-2, N-(2-Deoxy-α,β-D-glucopyranose-2-)-N²-acetyl-S-nitroso-D,L-penicillaminamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H23N3O8S
CAS Number:
Molecular Weight:
381.40
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

green

solubility

aqueous buffer: >100 mg/mL
methanol: soluble
water: soluble

shipped in

ambient

storage temp.

−20°C

SMILES string

CC(=O)NC(C(=O)NC1C(C(C(OC1O)CO)O)O)C(C)(C)SN=O

Related Categories

General description

A nitric oxide donor that exhibits greater stability than SNAP in aqueous medium, in both the presence and absence of EDTA. The half life of Glyco-SNAP-2 in aqueous medium is 27.4 h at room temperature, in the absence of EDTA, compared to 10.3 h for SNAP. The compound consists of a S-nitroso-N-acetylpencilamine moiety, which acts as a NO-donor, and a sugar fragment, which provides the desired hydrophobicity.
A water-soluble nitric oxide (NO) donor that exhibits greater stability (t½ = 27 hours) than that of SNAP. Exhibits properties similar to those of Glyco-SNAP-1 (Cat. No. 362338).

Biochem/physiol Actions

Cell permeable: no
Primary Target
Water-soluble nitric oxide (NO) donor
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Standard Handling (A)

Reconstitution

Unstable in solution; reconstitute just prior to use.

Other Notes

Hou, Y., et al. 1999. Methods Enzymol. 301, 242.
Ramirez, J., et al. 1996. Bioorg. Med. Chem. Lett.6, 2575.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service