Skip to Content
Merck
All Photos(1)

Key Documents

362330

Sigma-Aldrich

GAG Antagonist, Surfen

The GAG Antagonist, Surfen controls the biological activity of GAG. This small molecule/inhibitor is primarily used for Activators/Inducers applications.

Synonym(s):

GAG Antagonist, Surfen, Gβγ Activator, 12155, NSC12155, bis-2-Methyl-4-amino-quinolyl-6-carbamide, diHCl, trihydrate, Glycosaminoglycans Antagonist

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H20N6O · 2HCl · 3H2O
Molecular Weight:
499.39
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

white

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

A symmetrical quinolyl-urea compound that binds GAGs (glycosaminoglycans) via electrostatic interaction with the negatively charged sulfate and carboxyl moieties present in heparan sulfate (HS), heparin, and dermatan sulfate, resulting in effective blockage of GAGs interactions with their protein binding partners. Surfen is shown to effectively neutralize HS- and heparin-mediated thrombin inhibition of Factor Xa activity as well as heparin′s anti-clotting activity. Also reported to inhibit FGF2-induced Erk phosphorylation and tubulation in murine lung endothelial cultures (IC50 ~5 µM), fibronectin HS-binding domain-dependent CHO cell adhesion (IC50 = 3 µM), and HSV-1 infection of glucosaminyl 3-O-sulfotransferase-3A-expressing CHO cells (complete inhibition at 5 µM). Surfen analogs with improved potency may serve as promising candidates as less toxic alternatives to Protamine (Cat. No. 539122) in clinical applications.Reported to directly bind Gβγ subunit in a reversible manner, displace Gα-GDP from Gβγ, and acutely activate Gβγ signaling without Gα activation.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot into glass vials and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Surve, C.R., et al. 2016. Sci. Signal.9, ra22.
Schuksz, M., et al. 2008. Proc. Natl. Acad. Sci. USA105, 13074.
Hunter, D.T. Jr., and Hill, J.M. 1961. Nature191, 1378.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ferenc Zsila
Chirality, 27(9), 605-612 (2015-06-23)
Despite the diverse biological activities of the glycosaminoglycan (GAG) antagonist surfen, the molecular details of its interaction with biomacromolecules remain poorly understood. Therefore, heparin and DNA binding properties of surfen were studied by circular dichroism (CD) and UV absorption spectroscopy
Safder S Ganaie et al.
Cell, 184(20), 5163-5178 (2021-09-25)
Rift Valley fever virus (RVFV) is a zoonotic pathogen with pandemic potential. RVFV entry is mediated by the viral glycoprotein (Gn), but host entry factors remain poorly defined. Our genome-wide CRISPR screen identified low-density lipoprotein receptor-related protein 1 (mouse Lrp1/human
Ferenc Zsila
Biochemical and biophysical research communications, 460(3), 863-867 (2015-04-02)
It is shown that the heparin antagonist bis-aminoquinoline derivative surfen interacts with sulfated cyclodextrins in a unique fashion. Analysis of the UV spectroscopic data revealed exceptionally strong association (K(a) ∼ 10(7) M(-1)) of several surfen molecules to the external surface

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service