Skip to Content
Merck
All Photos(1)

Key Documents

344096

Sigma-Aldrich

Fluvastatin, Sodium Salt

InSolution, ≥98% inhibitor of HMG-CoA reductase

Synonym(s):

InSolution Fluvastatin, Sodium Salt, (±)-(3Rʹ,5Sʹ,6E)-7-(3-(4-Fluorophenyl)-1-isopropylindol-2-yl)-3,5-dihydroxy-6-heptenoate, sodium

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€136.00

€136.00


Please contact Customer Service for Availability


Select a Size

Change View
10 MG
€136.00

About This Item

Empirical Formula (Hill Notation):
C24H25FNNaO4
CAS Number:
Molecular Weight:
433.45
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€136.00


Please contact Customer Service for Availability

Quality Level

Assay

≥98% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles
protect from light

shipped in

dry ice

storage temp.

−70°C

SMILES string

[Na+].Fc1ccc(cc1)c2c3c([n](c2\C=C\[C@H](O)C[C@H](O)CC(=O)[O-])C(C)C)cccc3

InChI

1S/C24H26FNO4.Na/c1-15(2)26-21-6-4-3-5-20(21)24(16-7-9-17(25)10-8-16)22(26)12-11-18(27)13-19(28)14-23(29)30;/h3-12,15,18-19,27-28H,13-14H2,1-2H3,(H,29,30);/q;+1/p-1/b12-11+;/t18-,19-;/m0./s1

InChI key

ZGGHKIMDNBDHJB-RPQBTBOMSA-M

General description

A synthetic, competitive inhibitor of HMG-CoA reductase that acts as anti-hypercholesterolemic agent. Unlike Lovastatin (Cat. No. 438185), Mevastatin (Cat. No. 474700), and Simvastatin (Cat. No. 567020), it does not require activation prior to use for in vitro studies. Reduces the basal MMP-1 levels in culture media of endothelial cells in a time- and dose-dependent manner. Fluvastatin is metabolized in the liver, primarily via hydroxylation of the indole ring at the 5- and 6-positions. N-Dealkylation and β-oxidation of the side-chain also occurs. The hydroxy metabolites are also reported to have some pharmacologic activity.

Biochem/physiol Actions

Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Other Notes

Yamamoto, A., et al. 2001. J. Pharm. Pharmacol.53, 227.
Dansette, P.M., et al. 2000. Exp. Toxicol. Pathol.52, 145.
Ikeda, U., et al. 2000. Hypertension36, 325.
Levy, R.I., et al. 1993. Circulation87 (4 Suppl.), III45.
Tse, F.L.S. et al. 1992. J. Clin. Pharmacol.32, 630.
Yuan, J., et al. 1991. Atherosclerosis87, 147.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service