341612
InhibitorSelect FGF Signaling Pathway Inhibitor Panel
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352200
form
solid
Quality Level
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
desiccated (hygroscopic)
protect from light
solubility
DMSO: soluble (supplied)
shipped in
ambient
storage temp.
−20°C
General description
A panel containing 14 potent, selective, reversible and cell-permeable inhibitors useful for the study of multiple signaling pathways activated by FGF: 1 mg of Akt Inhibitor VIII, Isozyme-Selective, Akti-1/2 (Cat. No. 124018); 2 mg of EGF/FGF/PDGF Receptor Tyrosine Kinase Inhibitor (Cat. No. 324841); 5 mg of ET-18-OCH3 (Cat. No. 341207); 2 mg of FGF/VEGF Receptor Tyrosine Kinase Inhibitor, PD173074 (Cat. No. 341607); 5 mg of FGF Receptor Tyrosine Kinase Inhibitor (Cat. No. 341608); 500 µg of Go 6983 (Cat. No. 365251); 5 mg of JNK Inhibitor VIII (Cat. No. 420135); 5 mg of LY 294002 (Cat. No. 440202); 5 mg of PD 98059 (Cat. No. 513000); 1 mg of PP2 (Cat. No. 529573); 5 mg of Rac1 Inhibitor (Cat. No. 553502); 10 mg of STAT3 Inhibitor III, WP1066 (Cat. No. 573097); 1 mg of U0126 (Cat. No. 662005); 1 mg of ZM 336372 (Cat. No. 692000). Also provided with 15 ml of anhydrous DMSO (Cat. No. KP31817). Supplied with a data sheet.
A panel containing 15 potent and selective inhibitors useful for the study of FGF signaling pathway. This panel contains the following inhibitors and 15 ml anhydrous DMSO for reconstitution:
Packaging
Packaged under inert gas
Warning
Toxicity: Regulatory Review (Z)
Reconstitution
Following reconstitution, aliquot and freeze (-20°C) each inhibitor. Each inhibitor solution is stable for up to 3 months at -20°C.
Please refer to information provided on the CD for reconstitution information of individual inhibitors.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
10-13 - German Storage Class 10 to 13
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service