Skip to Content
Merck
All Photos(1)

Key Documents

324895

Sigma-Aldrich

eIF-2α Inhibitor, Salubrinal

eIF-2α Inhibitor, Salubrinal, CAS 304475-63-6, is a cell-permeable, selective inhibitor of the phosphatase complexes that dephosphorylate eIF-2α. Protects cells against ER stress-induced apoptosis.

Synonym(s):

eIF-2α Inhibitor, Salubrinal, Sal

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H17Cl3N4OS
CAS Number:
Molecular Weight:
479.81
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥90% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

beige

solubility

DMSO: 10 mg/mL

shipped in

wet ice

storage temp.

−20°C

General description

A cell-permeable and selective inhibitor of the phosphatase complexes that dephosphorylate eukaryotic translation initiation factor 2 subunit α (eIF-2α). Shown to offer protection against ER stress-induced apoptosis (EC50 ~15 µM in PC12 cells stimulated with 750 ng/ml of Tunicamycin (Cat. No. 654380). Prevents HSV replication both in Vero cells (IC50 ~3 µM for reducing plaque formation) in vitro and in a mouse cornea infection model in vivo.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
elF-2α
Product does not compete with ATP.
Reversible: no
Target IC50: 3 µM preventing HSV replication both in Vero cells.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Boyce, M., et al. 2005. Science307, 935.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ziwei Zhang et al.
Molecular nutrition & food research, 66(5), e2100791-e2100791 (2021-12-31)
To investigate the role of endoplasmic reticulum stress (ERS)-induced autophagy in inflammatory bowel disease (IBD) and the intervention mechanism of Portulaca oleracea L. (POL) extract, a medicinal herb with anti-inflammatory, antioxidant, immune-regulating, and antitumor properties, in vitro and in vivo.
YongYao Xu et al.
PloS one, 17(7), e0271656-e0271656 (2022-07-29)
Inflammatory signaling induces barrier dysfunction in retinal-pigmented epithelium (RPE) cells and plays a role in the pathology of age-related macular degeneration (AMD). We studied the role of Zn flux from the endoplasmic reticulum (ER) to the cytoplasm via Zip7 during
Ting Zhu et al.
Veterinary research, 52(1), 148-148 (2021-12-22)
Viruses have evolved multiple strategies to manipulate their host's translational machinery for the synthesis of viral proteins. A common viral target is the alpha subunit of eukaryotic initiation factor 2 (eIF2α). In this study, we show that global protein synthesis
Ai-Ling Tian et al.
Cell death & disease, 12(1), 6-6 (2021-01-09)
The integrated stress response manifests with the phosphorylation of eukaryotic initiation factor 2α (eIF2α) on serine residue 51 and plays a major role in the adaptation of cells to endoplasmic reticulum stress in the initiation of autophagy and in the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service