Skip to Content
Merck
All Photos(1)

Key Documents

2EC

Sigma-Aldrich

Enzyme Conjugate (100X)

Sign Into View Organizational & Contract Pricing

Select a Size

2EC
€61.20

€61.20


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
2EC
€61.20

About This Item

UNSPSC Code:
12352204
NACRES:
NA.84

€61.20


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service