Skip to Content
Merck
All Photos(1)

Key Documents

23-057-M

Sigma-Aldrich

Smac/DIABLO

Synonym(s):

Diablo homolog, mitochondrial, Direct IAP-binding protein with low pI, Second mitochondria-derived activator of caspase (Smac)

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€640.00
250 μG
€3,700.00

€640.00


Please contact Customer Service for Availability


Select a Size

Change View
10 μG
€640.00
250 μG
€3,700.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.32

€640.00


Please contact Customer Service for Availability

biological source

human

Quality Level

form

liquid

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable

NCBI accession no.

General description

Smac/DIABLO is a mitochondrial protein that is proteolytically processed and released during apoptosis along with cytochrome c and other pro-apoptotic factors. Once in the cytosol, Smac/DIABLO binds to inhibitors of apoptosis (IAP) proteins and disrupts the ability of the IAPs to inhibit caspases 3, 7, and 9 thereby promoting apoptosis. Along with several other pro-apoptotic proteins, Smac/DIABLO has been shown to be a substrate for the ubiquitin ligase XIAP.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service