Skip to Content
Merck
All Photos(1)

Documents

189488

Sigma-Aldrich

Autophagy Regulators Panel

A panel containing 23 cell-permeable, potent, and selective small molecule regulators that is useful for the study of autophagy process.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

shipped in

wet ice

storage temp.

−20°C

General description

A panel containing 23 cell-permeable, potent, and selective small molecule regulators that is useful for the study of autophagy process. The panel contains the molecules listed in the table and 15 ml of anhydrous DMSO.
A panel containing 23 cell-permeable, potent, and selective small molecule regulators that is useful for the study of autophagy process:10 mg of Leupeptin, Hemisulfate, Synthetic (Cat. No. 108976),1 mg of AMPK Inhibitor, Compound C (Cat. No. 171260),50 mg of Autophagy Inhibitor, 3-MA (Cat. No. 189490),10 mg of Autophagy Inducer, STF-62247 (Cat. No. 189497),10 µg of Bafilomycin A1, Streptomyces griseus (Cat. No. 196000),10 mg of bpV(phen) (Cat. No. 203695)10 mg of Dynamin Inhibitor I, Dynasore (Cat. No. 324410),10 mg of DAPK Inhibitor (Cat. No. 324788),1 mg of HDAC Inhibitor XXIII, Tubastatin A (Cat. No. 382187),5 mg of mTOR Inhibitor III, PP242 (Cat. No. 475988),5 mg of Necrostatin-1 (Cat. No. 480065),5 mg of Nigericin, Sodium Salt, Streptomyces hygroscopicus (Cat. No. 481990),10 mg of Nocodazole (Cat. No. 487928),10 mg of Pifithrin-μ (Cat. No. 506155),1 mg of PI-103 (Cat. No. 528100),1 mg of Rapamycin (Cat. No. 553210),5 mg of PIKfyve Inhibitor (Cat. No. 524611),10 mg of SMER28 (Cat. No. 573121),5 mg of Tamoxifen, 4-Hydroxy-, (Z)- (Cat. No. 579002),1 mg of Thapsigargin (Cat. No. 586005)1 mg of Caspase Inhibitor I, Z-VAD-fmk (Cat. No. 627610),and 10 mg of Tunicamycin, Streptomyces lysosuperficus (Cat. No. 654380), and1 mg of Wiskostatin (Cat. No. 681525).Also provided with 15 ml of anhydrous DMSO (Cat. No. KP31817). Supplied with a data sheet.

Also available separately: Spautin-1 (Cat. No. 567569)

Packaging

Packaged under inert gas

Warning

Toxicity: Multiple Toxicity Values, refer to MSDS (O)

Reconstitution

Please refer to information provided on the CD for reconstitution of individual inhibitors.

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.
Fleming, A., et al. 2011. Nat. Chem. Biol.7, 9.
Mizushima, N. and Levine, B., 2010. Nat. Cell Biol.12, 823.
Klionsky, D.J., et al. 2010. Autophagy6, 1017.
Mizushima, N., et al. 2010. Cell140, 313.
Zhang, L., et al. 2007. Proc. Natl. Acad. Sci. USA104, 19023.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

188.6 °F - (refers to pure substance)

Flash Point(C)

87 °C - (refers to pure substance)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service