Skip to Content
Merck
All Photos(1)

Key Documents

172060

Sigma-Aldrich

Andrographolide

A bicyclic diterpenoid lactone that displays anti-viral, anti-inflammatory, anti-apoptotic, and anti-hyperglycemic properties.

Synonym(s):

Andrographolide, Andro

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
€80.70

€80.70


Please contact Customer Service for Availability


Select a Size

Change View
50 MG
€80.70

About This Item

Empirical Formula (Hill Notation):
C20H30O5
CAS Number:
Molecular Weight:
350.45
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€80.70


Please contact Customer Service for Availability

Quality Level

description

Merck USA index - 14, 633

Assay

≥95% (TLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

chloroform: methanol (1:1): 10 mg/mL
DMSO: 15 mg/mL (Use only fresh DMSO.)

shipped in

ambient

storage temp.

2-8°C

SMILES string

O1CC(\C(=C/C[C@H]2[C@]3(C(C(C(CC3)O)(CO)C)CCC2=C)C)\C1=O)O

InChI

1S/C20H30O5/c1-12-4-7-16-19(2,9-8-17(23)20(16,3)11-21)14(12)6-5-13-15(22)10-25-18(13)24/h5,14-17,21-23H,1,4,6-11H2,2-3H3/b13-5+/t14-,15?,16?,17?,19+,20?/m1/s1

InChI key

BOJKULTULYSRAS-ZJFCSBQFSA-N

General description

A bicyclic diterpenoid lactone that displays anti-viral, anti-inflammatory, anti-apoptotic, and anti-hyperglycemic properties. Acts as an irreversible antagonist of NF-κB and AP-1 (IC50 ≤ 15 µM) activation, and prevents in vitro and in vivo T cell activation. Exerts no effect on IκBα degradation, p50 and p65 nuclear translocation. Inhibits iNOS and Mac-1 expressions and ROS production, and prevents endotoxin shock in mouse model. Further, reported to activate PI 3K/Akt pathway.
An irreversible blocker of NF-κB and AP-1 (IC50 ≤ 15 µM) activation that displays anti-viral, anti-inflammatory, anti-apoptotic, and anti-hyperglycemic properties. Exerts no effect on IκBα degradation of nuclear translocation p50 and p65. Inhibits iNOS and Mac-1 expressions and ROS production, and prevents endotoxin shock in a mouse model. Reported to supress Cytochrome C release and apoptosis by activating the PI 3-K/Akt pathway.

Biochem/physiol Actions

Cell permeable: no
Primary Target
NF-κB and AP-1 activation
Product does not compete with ATP.
Reversible: no
Target IC50: ≤ 15 µM against NF-κB and AP-1 activation

Packaging

Packaged under inert gas

Warning

Toxicity: Carcinogenic / Teratogenic (D)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Xia, Y.F., et al. 2004. J. Immunol.173, 4207.
Chen, J.H., et al. 2004. Biochem. Pharmacol.67, 1337.
Yu, B.C., et al. 2003. Planta Med.69, 1075.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service