Skip to Content
Merck
All Photos(1)

Key Documents

166904

Millipore

Tryptic Soy Broth

bottle of, ready-to-use, bottle volume 100 mL , filling volume

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
Pricing and availability is not currently available.

Agency

EP 2.6.1
JP 4.06
USP 71

Quality Level

sterility

sterile; autoclaved

form

liquid

feature

closure type (screw cap with septum)
ready-to-use

packaging

bottle of

technique(s)

sterile filtration: suitable

bottle capacity

125 mL

bottle volume

100 mL , filling volume

application(s)

cosmetics
food and beverages
pharmaceutical
sterility testing

storage temp.

2-25°C

suitability

nonselective for

General description

Tryptic SoyBroth is a complex nutrient rich inhibitor free microbial culture mediadesigned in compliance with European (EP), Japanese (JP) and United StatesPharmacopeia (USP).The medium issupplemented with peptones from casein and soymeal to provide an enhancedcarbon source, that promotes the growth of the microbes present in the testsample.

Application

Tryptic Soy Broth is used for the isolation andcultivation of fastidious bacteria, yeast. TSB is suitable for sterilitytesting of substances, preparations, and products from pharmaceutical and foodindustry.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service