Skip to Content
Merck
All Photos(1)

Key Documents

14-931-M

Sigma-Aldrich

CaMKK2, active

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€508.00
250 μG
€3,340.00

€508.00


Please contact Customer Service for Availability


Select a Size

Change View
10 μG
€508.00
250 μG
€3,340.00

About This Item

UNSPSC Code:
12352200
eCl@ss:
32160405
NACRES:
NA.41

€508.00


Please contact Customer Service for Availability

biological source

human

Quality Level

mol wt

Mw 64 kDa

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

NCBI accession no.

UniProt accession no.

Gene Information

human ... CAMKK2(10645)

General description

Calcium/calmodulin-dependent protein kinase belonging to a proposed calcium-triggered signaling cascade involved in a number of cellular processes. Isoform 1, isoform 2 and isoform 3 phosphorylate CAMK1 and CAMK4. Isoform 3 phosphorylates CAMK1D. Isoform 4, isoform 5 and isoform 6 lacking part of the calmodulin-binding domain are inactive. Efficiently phosphorylates 5′-AMP-activated protein kinase (AMPK) trimer, including that consisting of PRKAA1, PRKAB1 and PRKAG1. This phosphorylation is stimulated in response to Ca2+ signals. Seems to be involved in hippocampal activation of CREB1. May play a role in neurite growth. Isoform 3 may promote neurite elongation, while isoform 1 may promoter neurite branching.
Product Source: N-terminal 6His-tagged, recombinant, human CaMKK2 full length, expressed by baculovirus in Sf21 insect cells

Packaging

Also available in 250μg size as catalog # 14-931M and in bulk as catalog # 14-931-K.

Quality

Routinely evaluated by phosphorylation of LSNLYHQGKFLQTFCGSPLYRRR

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service