Skip to Content
Merck
All Photos(1)

Key Documents

1.46817

Millipore

IsoBag® TSA LT Contact+

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
€1,590.00

€1,590.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EACH
€1,590.00

About This Item

UNSPSC Code:
41106231

€1,590.00


Please contact Customer Service for Availability

sterility

sterile; γ-irradiated

Quality Level

form

plate

manufacturer/tradename

IsoBag®

application(s)

pharmaceutical

compatibility

for use with Surface monitoring

General description

The IsoBag® TSA LT contact+ is designed for quick and convenient transfer of culture media for surface monitoring into isolators or RABS systems containing Getinge La Calhène DPTE Alpha Ports of 190 mm diameters.

Application

Environmental monitoring for isolators (surface monitoring). Detection of fungi and bacteria

Legal Information

ISOBAG is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service