1.46572
Thioglycollate Broth with Tween®80
bottle volume 850 mL , filling volume
Synonym(s):
FTM, Thioglycollate Broth
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Agency
EP 2.6.13
JP 4.06
USP 71
Quality Level
Sterilization Compatibility
autoclavable compatible
form
liquid
feature
closure type screw cap with septum
packaging
pack of 12 units
bottle of
bottle capacity
1000 mL
bottle volume
850 mL , filling volume
pH
6.9
application(s)
food and beverages
pharmaceutical
sterility testing
components
tween
storage temp.
2-25°C
suitability
nonselective for
General description
Thioglycollate Broth with Tween®80 is a complex medium designed for the cultivation and isolation of fastidious aerobic and anaerobic microorganisms present in sterility test samples of pharmaceutical industry.
The anaerobic atmosphere is created through reducing the redox potential of the medium by Thioglycollate and L-Cystine. Additionally, these compounds inactivate mercury and other heavy metal elements present in the environment.
Tween®80 is effective against quaternary ammonium compounds, amphoteric surfactants, benzamidines, chlorhexidines, and dequadin. Tween®80 inactivates benzyl alcohol, carbanilides, di-chlorobenzyl alcohols, benzoic acid, p-hydroxybenzoic acid and its esters, phenols, phenylethyl alcohols, and Solbrol (PHB esters).
The anaerobic atmosphere is created through reducing the redox potential of the medium by Thioglycollate and L-Cystine. Additionally, these compounds inactivate mercury and other heavy metal elements present in the environment.
Tween®80 is effective against quaternary ammonium compounds, amphoteric surfactants, benzamidines, chlorhexidines, and dequadin. Tween®80 inactivates benzyl alcohol, carbanilides, di-chlorobenzyl alcohols, benzoic acid, p-hydroxybenzoic acid and its esters, phenols, phenylethyl alcohols, and Solbrol (PHB esters).
Application
Thioglycollate Broth with Tween®80 is used as a sterility testing medium for sterile pharmaceutical samples. FTM clear is mainly used to cultivate and isolate anaerobic and microaerophilic microbes.
Legal Information
TWEEN is a registered trademark of Croda International PLC
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service