Skip to Content
Merck
All Photos(1)

Key Documents

14-642-M

Sigma-Aldrich

PTPMEG-1 Protein, 10 µg

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€436.00
250 μG
€3,340.00

€436.00


Please contact Customer Service for Availability


Select a Size

Change View
10 μG
€436.00
250 μG
€3,340.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.32

€436.00


Please contact Customer Service for Availability

biological source

human

Quality Level

100
300

mol wt

Mw 83.1 kDa

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (phosphatase)

NCBI accession no.

UniProt accession no.

General description

N-Terminal GST-tagged, recombinant, human PTPMEG-1, amino acids 423-end
recombinant human PTPMEG-1, amino acids 423-end

Application

N-terminal GST-tagged, recombinant, human PTPMEG-1, amino acids 423-end, for use in Phosphatase Assays.
Research Category
Inflammation & Immunology

Biochem/physiol Actions

  • Protein Target: PTPMEG-1
  • Target Sub-Family:Tyr phosphatase

Quality

routinely evaluated in a PTPMEG-1 Phosphatase Assay

Physical form

Format: Glutathione agarose affinity chromatography

Storage and Stability

6 months at -70°C

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service