Skip to Content
Merck
All Photos(1)

Key Documents

14-328

Sigma-Aldrich

JNK1α1/SAPK1c Protein, inactive, 50 g

Unactive, N-terminal His6-tagged recombinant, human, full-length JNK1α1/SAPK1c, for use in Kinase Assays.

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€545.00

€545.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
50 μG
€545.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.32

€545.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

human

Quality Level

recombinant

expressed in baculovirus infected Sf21 cells

mol wt

Mw 45 kDa

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

UniProt accession no.

shipped in

dry ice

Gene Information

human ... MAPK8(5599)

General description

N-terminal His6-tagged recombinant, human, full-length JNK1α1/SAPK1c
Product Source: Expressed by baculovirus in Sf21 insect cells

Biochem/physiol Actions

Protein Target: JNK1α1/SAPK1c
Target Sub-Family: CMGC

Quality

Routinely evaluated by phosphorylation of ATF2 (amino acids 19-196)

Physical form

Ni2+/NTA-agarose

Storage and Stability

6 months at -20°C

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V I Peinado et al.
Avian pathology : journal of the W.V.P.A, 21(1), 55-64 (1992-01-01)
Reference values for some haematological and plasma chemical values in four species of clinically normal adult flamingos were established for use in avian medicine. The following variables were studied in rosy, greater, Chilean and lesser flamingos: haematocrit, haemoglobin concentration, erythrocyte
Anna F Lee et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 24(41), 9174-9184 (2004-10-16)
The p53 family member, p73, is essential for the survival of sympathetic neurons during the developmental period of naturally occurring neuronal death. Here, we have asked whether DeltaNp73, which is the only p73 isoform expressed in sympathetic neurons, mediates this
S Gupta et al.
Science (New York, N.Y.), 267(5196), 389-393 (1995-01-20)
Treatment of cells with pro-inflammatory cytokines or ultraviolet radiation causes activation of the c-Jun NH2-terminal protein kinase (JNK). Activating transcription factor-2 (ATF2) was found to be a target of the JNK signal transduction pathway. ATF2 was phosphorylated by JNK on

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service