Skip to Content
Merck
All Photos(1)

Key Documents

14-249-M

Sigma-Aldrich

p38δ/SAPK4 Protein, active, 10 µg

Active, recombinant GST fusion protein fused to full length human p38δ/SAPK4. For use in Kinase Assays.

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€579.00
250 μG
€3,770.00

€579.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 μG
€579.00
250 μG
€3,770.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.41

€579.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

human

Quality Level

recombinant

expressed in E. coli

mol wt

Mw 71 kDa

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

NCBI accession no.

UniProt accession no.

General description

Product Source: expressed in E. coli
Recombinant GST fusion protein fused to full length human p38δ/SAPK4

CHK3

Biochem/physiol Actions

Protein Target: p38δ/SAPK4
Target Sub-Family: CMGC

Quality

Routinely evaluated by phosphorylation of MBP in vitro

Physical form

Affinity chromatography on glutathione Sepharose beads after activation with SKK3/MEK6

Storage and Stability

6 months at -20°C

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

Sepharose is a trademark of Cytiva
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Goedert et al.
The EMBO journal, 16(12), 3563-3571 (1997-06-01)
A cDNA was cloned that encodes human stress-activated protein kinase-4 (SAPK4), a novel MAP kinase family member whose amino acid sequence is approximately 60% identical to that of the other three SAP kinases which contain a TGY motif in their

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service