Skip to Content
Merck

1.37009

SAFC

Potassium chloride

EMPROVE® EXPERT, Ph. Eur., BP, JP, USP

Pharma Manufacturing

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KCl
CAS Number:
Molecular Weight:
74.55
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-211-8

Agency

BP
JP
Ph. Eur.
USP

Quality Level

description

EMPROVE® EXPERT Ph Eur,BP,USP,JP

product line

EMPROVE® EXPERT

form

solid

feature

RNase and DNase free

pH

5.5-8.0 (25 °C, 50 g/L in H2O)

bp

1413 °C/1013 hPa

mp

770 °C (lit.)

density

1.98 g/mL at 25 °C (lit.)

application(s)

liquid formulation
ophthalmics
parenterals
pharma/biopharma processes
pharmaceutical

storage temp.

15-25°C

SMILES string

[Cl-].[K+]

InChI

1S/ClH.K/h1H;/q;+1/p-1

InChI key

WCUXLLCKKVVCTQ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Finding the right excipient that matches your needs as well as regulatory demands can be a complicated challenge in formulation. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Potassium chloride is a mineral salt which is used for a broad range of applications in pharmaceutical and food industry. Pharmaceutical applications are e.g. treatment of hypokalemia, or serving as a component of isotonic solutions for infusion. Beyond, Potassium chloride can also serve as a strengthening agent and flavor enhancer for food and beverage.
Due to its low microbial and endotoxin limits, Potassium chloride Emprove® Expert is especially suitable for high-risk applications.
May be used for the preparation of phosphate buffered saline, and for the extraction and solubilization of proteins.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service