Skip to Content
Merck
All Photos(1)

Key Documents

1.25090

Millipore

Fractogel® EMD SO3 ⁻ (M) Resin

RoboColumn Fractogel® SO3 (M), volume 0.2 mL, prepacked column

Synonym(s):

RoboColumn Fractogel® SO3 (M), 0.2 ml

Sign Into View Organizational & Contract Pricing

Select a Size

8 PKG
€291.00

€291.00


Please contact Customer Service for Availability


Select a Size

Change View
8 PKG
€291.00

About This Item

UNSPSC Code:
41115711

€291.00


Please contact Customer Service for Availability

ligand

(Sulfoisobutyl)

material

polypropylene housing

product line

Fractogel®

form

prepacked column

parameter

≤8 bar pressure
~200 cm/hr max. flow rate

column I.D. × L

5 mm × 10 mm

volume

0.2 mL

matrix active group

Methacrylate

mean particle size

40-90 μm

capacity

~130 mL, resin binding capacity

application(s)

mAb
recombinant protein
vaccine development

separation technique

cation exchange

storage temp.

room temp

General description

5 x 10 mm

Features and Benefits

RoboColumn Fractogel® SO3 (M), 0.2 ml enables:
  • Increased binding strength at low pH
  • High aggregate removal efficiency combined with high dynamic binding capacity
  • Lower elution volumes for the highest purity levels
  • Compatibility with 2.5 % (v/v) aqueous benzyl alcohol containing 150 mM NaCl storage solution


Due to the titration behavior, the ion exchange capacity can be used from pH 2 to pH 12. The separation of proteins is based on reversible electrostatic interactions between the surface′s positively charged regions and the support. Proteins are retained efficiently on Fractogel® EMD SO3- (M) when the pH of the buffer is about 1 unit below their isoelectric points (pl).

The strength of the binding depends on the following:
  • the buffer system
  • pH value of the buffer which determines the surface charge of the protein
  • the concentration of the counter ions
  • the charge density on the support (protein binding capacity)

Legal Information

FRACTOGEL is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service