Skip to Content
Merck
All Photos(1)

Documents

1.24511

Millipore

1,4-Dithioerythritol

for biochemistry

Synonym(s):

1,4-Dithioerythritol, Clelands reagent, 1,4-Dimercapto-2,3-butanediol, DTE

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H10O2S2
CAS Number:
Molecular Weight:
154.25
MDL number:
UNSPSC Code:
12352104
EC Index Number:
229-998-8
NACRES:
NA.21

Quality Level

Assay

≥98.0%

form

lyophilized

reaction suitability

reagent type: reductant

mp

83 °C

storage temp.

2-8°C

InChI

1S/C4H10O2S2/c5-3(1-7)4(6)2-8/h3-8H,1-2H2/t3-,4+

InChI key

VHJLVAABSRFDPM-ZXZARUISSA-N

Analysis Note

Assay (iodometric): ≥ 99 %
Identity (IR-spectrum): passes test
Melting point: 81 - 85 °C
Heavy metals (as Pb): ≤ 0.001 %
Fe (Iron): ≤ 0.0005 %

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service