Skip to Content
Merck
All Photos(1)

Documents

119134

Sigma-Aldrich

Adenosine 5′-O-(2-Thiodiphosphate), Trilithium Salt

A P₂-purinergic receptor agonist that acts as a potent insulin secretagogue.

Synonym(s):

Adenosine 5′-O-(2-Thiodiphosphate), Trilithium Salt, ADP-β-S, 3Li

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥80% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

impurities

≤10% ADP

color

white

solubility

water: 20 mg/mL

shipped in

wet ice

storage temp.

−20°C

InChI

1S/C10H15N5O9P2S/c11-8-5-9(13-2-12-8)15(3-14-5)10-7(17)6(16)4(23-10)1-22-25(18,19)24-26(20,21)27/h2-4,6-7,10,16-17H,1H2,(H,18,19)(H2,11,12,13)(H2,20,21,27)/t4-,6-,7-,10-/m1/s1

InChI key

HCIKUKNAJRJFOW-KQYNXXCUSA-N

General description

A P2-purinergic receptor agonist that acts as a potent insulin secretagogue. Substrate and inhibitor of ADP-dependent enzyme systems.
Substrate and inhibitor of ADP-dependent enzyme systems. A P2-purinergic agonist that acts as a potent insulin secretagogue.

Biochem/physiol Actions

Cell permeable: no
Primary Target
A P2-purinergic receptor agonist
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Shalev, M., et al. 1999. J. Urol. 161, 955.
Hillaire-Buys, D., et al. 1993. Br. J. Pharmacol. 109, 183.
Bertrand, G., et al. 1991. Br. J. Pharmacol.102, 627.
Eckstein, F., and Goody, R.S. 1976. Biochemistry15, 1685.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service