Skip to Content
Merck
All Photos(1)

Key Documents

1.14964

Supelco

Software

configured for Windows, Spectroquant®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115311

product line

Spectroquant®

technique(s)

photometry: suitable

compatibility

configured for Windows
for use with Spectroquant® Nova 30
for use with Spectroquant® Nova 400
for use with Spectroquant® Nova 60 A

storage temp.

2-30°C

General description

for Windows (in English and German) for the data transfer of NOVA photometers Spectroquant®

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service