1.09215
Ziehl-Neelsen carbol-fuchsin solution
for microscopy
Synonym(s):
Carbol fuchsin solution
About This Item
Recommended Products
Quality Level
form
liquid
IVD
for in vitro diagnostic use
transition temp
flash point 47 °C
density
0.99 g/cm3 at 20 °C
application(s)
clinical testing
diagnostic assay manufacturing
hematology
histology
storage temp.
15-25°C
Related Categories
General description
The 500 ml bottle of Ziehl-Neelsen carbolfuchsin solution provides approx. 250 stainings. Pretreatment of the specimens with Sputofluol™ dissolves the bacteria from the surrounding viscid sputum and cell material. Ziehl-Neelsen carbolfuchsin solution - for microscopy is an IVD product and CE registered. For more details, please see instructions for use (IFU). The IFU can be downloaded from this webpage.
Application
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Flam. Liq. 3 - Muta. 2 - Skin Corr. 1B
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
116.6 °F
Flash Point(C)
47 °C
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service